Learn More
Eleutheroside E, MedChemExpress
Product Code. p-200099468
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16489036
Brand: Medchem Express HYN0272
MedChemExpress Eleutheroside E, a principal component of Eleutherococcus enticosus, has anti-inflammatory and protective effects in ischemia heart.
Eleutheroside E, a principal component of Eleutherococcus enticosus, has anti-inflammatory and protective effects in ischemia heart.
Specifications
Chemical Name or Material Eleutheroside E
CAS 39432-56-9
Color White
Purity Grade Notes Research
Molecular Formula C34H46O18
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (134.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.35 mM; Need ultrasonic)
SMILES COC(C=C1[C@H]2[C@@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C(OC)=C4)([H])[C@]3([H])CO2)=C(C(OC)=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO
Molecular Weight (g/mol) 742.72
Formula Weight 742.72
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only