Learn More
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
missing translation for 'unitSize'
Each
Description
Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With | EM/EME Series Heating Mantles |
| Voltage | 230 V |
| Wattage | 200w |
| Product Type | Heating Element |
| Frequency | 50/60 Hz |