Learn More
BTX™ Electrofusion Chambers - Meander Fusion Chambers
Click to view available options
Connection Type:
Banana Plug to Micrograbber
Unit Size:
Pack of 4
Description
- Novel microslide design which is specifically used for electro cell fusion
- Conductive metal alloy with two primary bars spaced 0.2 mm apart
- Designed for direct viewing of dimer formation
- Compatible with BTX Systems such as ECM 2001+ and Gemini X2
Specifications
Specifications
| For Use With (Application) | Cell Fusion, Hybridoma Production, Plant Protoplast Fusion |
| For Use With (Equipment) | ECM 2001+, Legacy ECM 2001 Electrofusion Generators |
| Material | Metal Alloy |
| Product Line | BTX Electrofusion |
| Product Type | Electrofusion accessory |
| Voltage | 16 VAC, 480 VDC |
| Description | Meander Fusion Chamber, 4 Pk. |
| Field Type | Divergent |
| Includes | 4 x Meander Fusion Chambers |
| Pulse Number Range | 1 to 99 (depending on voltage) |
| Show More |