Learn More
BINDER™ Electric Access Port for Series CB, CBF
BINDER™ Electric access port is an option for use with CO2 incubators CB and CBF.
Brand: BINDER™ 8612-0033
This item is not returnable.
View return policy
Description
- Electric access port
- 8-pin
- For low voltage with LEMO socket (coverable) and LEMO plug (max. 24 V – 2 A)
Specifications
Electric access port - 8-pin, for low voltage with LEMO socket (coverable) and LEMO plug (max. 24 V - 2 A) | |
BINDER Chambers Series CB, CBF | |
Access Port | |
Low Voltage with LEMO Socket (Coverable) and LEMO Plug (Max. 24 V - 2 A) |
Product Content Correction
Product Title
BINDER™ Electric Access Port for Series CB, CBF > For Use With: BINDER Chambers Series CB, CBF