Learn More
Tocris Bioscience™ EED 226
Potent and selective EED inhibitor
Brand:  Tocris Bioscience™ 7762/50
548.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18719266
  • € 694.00 / 50mg
Explore more special offers
Description
Description
EED 226 is a potent (IC50 = 23.4 nM) and selective EED inhibitor. It inhibits PRC2 activity by binding to the H3K27me3 pocket of EED. The compound shows selectivity for PRC2 complex over protein methyltransferases, kinases, and other protein classes. In vitro, EED 226 regulates histone H3K27 methylation and PRC2 target gene expression. In vivo, it induces tumor regression in a mouse xenograft model. EED 226 is orally bioavailable.
Specifications
Specifications
N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine
50 mg
C17H15N5O3S
2083627-02-3
EED