Learn More
Justrite™ EcoPolyBlend™ Accumulation Center
Product Code. p-7070908
Click to view available options
Holds:
6 Drums
8 Drums
Color:
Black
Unit Size:
Each
Product Code. 10629790
Brand: Justrite™ 28659
Only null left
Add to basket
Add to basket
Create custom systems to deliver a complete solution. The Justrite™ EcoPolyBlend™ Accumulation Center combines units to create customized spill-control systems, allowing for the perfect solution to specific industry needs.
Low-Profile EcoPolyBlend™ Accumulation Centers only measure 5.5 in. (140-mm) high for easy handling of drums. Made of recycled polyethylene, these accumulation centers are nearly impervious to chemicals, including acids and corrosives. Seamless construction eliminates leaks, making these ideal for drum filling and dispensing operations. Self-locating grates won’t ride-up, and are removable for easy cleaning. Combine 2-, 4-, 6-, and 8-drum modular units to create your own customized system. Use the optional joining clips to firmly lock units together. Sumps remain separate and clips can be removed for easy repositioning.
Specifications
Color Black
Holds 6 Drums