Learn More
Sharpsguard™ Eco Anatomical Waste Container
Click to view available options
Capacity (Metric):
2.6 L
10.91 L
21.01 L
Unit Size:
Pack of 10
Pack of 20
Pack of 48
Description
- Sharpsguard™ eco anatomical is for the safe disposal of anatomical and other potentially infectious waste.
Specifications
Specifications
| Capacity (Metric) | 10.91 L |
| Closure Color | Red |
| Diameter (Metric) Outer | 274 mm |
| Color | Yellow |
| Height (Metric) | 248 mm |
| Quantity | 20/Pk. |
| For Use With (Application) | Safe Disposal of Anatomical and Potentially Infectious Waste |