Learn More
Tocris Bioscience™ EC 144
High affinity, potent and selective Hsp90 inhibitor
Brand: Tocris Bioscience™ 4701/10
275.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
EC 144 is a high affinity, potent and selective Hsp90 inhibitor (Ki = 0.2 nM and IC50 = 1.1 nM). Exhibits selectivity for Hsp90 over Grp94 and TRAP1 (Ki values are 61 and 255 nM respectively) and has no effect (IC50 >10 μM) against a panel of 285 kinases. Degrades Her-2 in MCF-7 breast cancer cells (EC50 = 14 nM). Blocks tumor growth and induces partial tumor regression in an N87 gastric tumor mouse model. Also inhibits LPS-induced TNFα release in an LPS shock mouse model and suppresses disease development in a rat model of collagen-induced arthritis. Exhibits <20-fold efficacy over BIIB 021 (Cat. No. 4608) in mice. Orally available and brain penetrant.Specifications
>98% | |
EC 144 | |
EC 144 |
10 mg | |
413.9 |
For Research Use Only