Learn More
Ebro Electronic™ EBI 20 Data Logger Series
Click to view available options
Alarm Types:
Flashing LCD Segment
LED Indicator
Channels:
1
2
Data Memory:
20,000/Channel
40,000 Readings
IP Rating:
IP52
IP67
Unit Size:
Each
Description
EBI 20 data loggers are very robust and are in use all over the world in transportation and for warehouse monitoring.
Specifications
Specifications
| Type | Datalogger |
| Display Type | LCD |
| Data Memory | 40,000 Readings |
| Calibration | Calibrated |
| Alarm Types | Flashing LCD Segment |
| Battery Type | Lithium |
| Battery Life | >24 Months |
| Channels | 1 |
| Certifications/Compliance | Factory Calibration certificate |
| IP Rating | IP67 |
| Show More |