Learn More
Abeba™ Dynamic 37335 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37335/41
 View more versions of this product
Product Code. 15660938
  • € 73.95 / 1 pair
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • White/black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Seamless upper
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
37335
White
41
Polyurethane