Learn More
Abeba™ Dynamic 37310 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 37310/47
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
37310 |
White | |
47 | |
Polyurethane |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Abeba™ Dynamic 37310 Shoes > Size: 47