Learn More
Applied Biosystems™ Dynabeads™ L10 Tubes
Dynabeads L10 Tubes
Brand:  Applied Biosystems™ 74003
114.82 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10139012
  • € 124.00 / Pack of 5
Explore more special offers
Description
Description
Flat-sided reusable glass tubes necessary for optimal recovery of cysts/oocysts during magnetic separation with the MPC-1 or MPC-6 magnetic particle concentrators when using Dynabeads™ anti-Cryptosporidium or Dynabeads™ GC-Combo.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass
Dynabeads™ MPC™
5 Tubes
Collection Tube
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3128607 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number74003
Lot Number2794186 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number74003
Lot Number2793262 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number74003
Lot Number2793712 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number74003
Lot Number2879100 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number74003
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ Dynabeads™ L10 Tubes > 5 tubes