Learn More
Berkshire™ Durx™ 770 Cleanroom wipe
Durx™ 770 is ideal for critical small parts cleaning. Durx™ 770 is enhanced with an ultra-low particle and fiber treatment to significantly reduce particles and fibers compared to Durx™ 670.
Brand: Berkshire™ DR770070712
Description
Binder treatment reduces particle and fiber generation. It can be used for cleaning and maintenance in laboratories, aerospace, electronics, food processing, photo processing, and semiconductor, circuit board, pharmaceutical, and biomedical device manufacture.Specifications
Polyester Cellulose | |
Knife-Cut | |
ISO Class 5-8 |
Nonwoven | |
18 x 18 cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0826679 | Certificate of Compliance | 13/09/2021 | DR770070712 |