Learn More
B Braun™ Durogrip™ Needle Holders
An instrument for guiding needles, suturing and gripping fine or course tissues.
€ 160.00 - € 168.00
Specifications
Product Type | Needle holders |
---|---|
Description | Serrated jaws |
For Use With (Application) | Guiding/holding needles and sutures |
Description
- Serrated jaws on both varieties
- Jaws with tungsten carbide inserts
- Safe hold on needle
Specifications
Needle holders | |
Serrated jaws | |
Guiding/holding needles and sutures |
Product Title
B Braun™ Durogrip™ Needle Holders