Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
14 mm
Unit Size:
Pack of 125
Pack of 250
Pack of 500
Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
- Hose olives enable the connection of hoses to laboratory apparatuses
- Glass articles for further processing
Specifications
Specifications
| Quantity | 500/Pk. |
| Item Description | For attaching to various apparatuses; Installation in different apparatus possible |
| Joint Size | 8 mm |