Learn More
DWK Life Sciences DURAN™ Standard Socket, finished
Product Code. p-7189389 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
8 mm
10 mm
13 mm
15 mm
17 mm
22 mm
28 mm
33 mm
40 mm
46 mm
50 mm
54 mm
58 mm
65 mm
75 mm
90 mm
110 mm
Unit Size:
Each
Pack of 10
Pack of 5
Pack of 50
Product Code. 12028184
Brand: DWK Life Sciences 286550001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For a rigid, self-supporting connection.
The conical ground joint is the most common joint in glass apparatus engineering. The DURAN™ sleeves, ground, comply with DIN 12249 Form E in their design and meet higher requirements for the surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • According to DIN 12249 Form E
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts
  • Minimized grease requirement
  • Very low leakage rate
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Pk.
Item Description Installation in glass apparatus and devices
Type Finished
Joint Size 33 mm
Thickness (Metric) Wall 2 mm