Learn More
SI Analytics™ DURAN™ Moisture Trap
Shop All SI Analytics Products
Click to view available options
Size:
NS 14
Unit Size:
Each
Description
- Colorless
- Has a neck size of 14
- For use with bottle top TZ 1508
Specifications
Specifications
| Color | Colorless |
| Type | Moisture Trap |
| Size | NS 14 |