Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
Brand:  Lenz Laborglasinstrumente™ 03002973
 View more versions of this product
Product Code. 10589611
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from Duran™ glass
  • Complies with DIN EN ISO 4797 standards
  • Includes conical ground joint
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass, Duran Glass
NS 29/32
166 mm
2000 mL
Medium
Round
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers