Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable
Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
HA 10
HB 10
HB 16
Type:
Ground Joint
Polished
Unit Size:
Pack of 10
Pack of 50
Description
DURAN™ KPG™ bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs: HA: without grease cup HB: with grease cup
- Available as ground or polished version in the following designs: HA: without grease cup HB: with grease cup
- Small gap dimensions allow a completely smooth running and working under vacuum.
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | As component for stirrer guides / stirrer coolers |
| Type | Ground Joint |
| Joint Size | HB 16 |