Learn More
DWK Life Sciences DURAN™ Jacketed Coil Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
Alle ansehen DWK Life Sciences Produkte
missing translation for 'orderingAttributeHoverText'
Standard Taper Joint No:
NS 24/29
NS 29/32
Length (Metric):
250 mm
400 mm
missing translation for 'unitSize'
Each
Beschreibung
Combining the cooling properties of the Liebig condenser with those of the Dimroth condenser produces the intensive cooler. This is characterized by its large cooling surface (cooling coil and outer jacket) and is therefore suitable for condensing vapors from reaction mixtures with low boiling points (distillation).
- This type of condenser has a very large heat exchange surface due to its coil condenser and double jacket and is thus especially suited for working with low boiling point media
- Two removable plastic olives for inlet and outlet of the cooling medium
- Tight closure due to standard ground joint
Spezifikation
Spezifikation
| For Use With (Application) | use as a reflux condenser for condensation and feedback of the (solvent) vapor to the reaction mixture; Distillation; All condensers are special sizes and not according to DIN |
| Length (Metric) | 400 mm |
| Quantity | 1 Ea. |
| Standard Taper Joint No | NS 29/32 |
| Material | Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |
| Type | Non-DIN Size |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur