Learn More
DWK Life Sciences DURAN™ Glass Stopcock for Aspirator Bottles, Standard Taper Ground Cone
Stopcock with glass key and a ground cone for Aspirator bottles
€ 90.40
Specifications
For Use With (Application) | Controlled dispensing of liquids and reagents |
---|---|
Material | Borosilicate Glass (3.3) |
No. per Pack | 1/Pk. |
Autoclavable | Not Autoclavable |
Description
Aspirator stopcock with glass key and a ground cone to fit the socket side-arm of DURAN™ aspirator bottles.
- All glass construction
- Tight closure due to standard ground joint
Specifications
Controlled dispensing of liquids and reagents | |
Borosilicate Glass (3.3) | |
1/Pk. | |
Not Autoclavable |
Product Title
DWK Life Sciences DURAN™ Glass Stopcock for Aspirator Bottles, Standard Taper Ground Cone