Learn More
Witeg™ Duran Glass Separatory Funnels
Witeg™ Duran™ Glass Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Brand: Witeg™ 2 677 200
This item is not returnable.
View return policy
Description
- Conical shape
- Graduated
- Economy-needle valve stopcock
- PTFE-needle valve
- Screw-thread and cap
Specifications
2000 mL | |
Duran Glass, Borosilicate Glass | |
1Pack | |
Separatory Funnel |