Learn More
Witeg™ Duran Glass Separatory Funnels
Witeg™ Duran™ Glass Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Witeg™ 2 677 100
 View more versions of this product
Product Code. 10632563
  • € 128.00 / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical shape
  • Graduated
  • Economy-needle valve stopcock
  • PTFE-needle valve
  • Screw-thread and cap
Specifications
Specifications
1000 mL
Duran Glass, Borosilicate Glass
1Pack
Separatory Funnel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers