Learn More
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints
Provides for secure and easy connection from a large cone to a small socket in glass tubing. Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01501419
This item is not returnable.
View return policy
Description
- Acc. to DIN 12257
- High taper accuracy of the cones
- Minimized need for grease
- Excellent tightness
- Corrosion-free and resistant to thermal shock
Specifications
DIN 12257 and in addition to DIN standard | |
Duran Glass | |
1Each |
NS 19/26 | |
Reduction Adapter |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints > Socket Size/Cone Size: NS 14/23/NS 19/26