Learn More
Lenz Laborglasinstrumente™ Expansion Adapter with Ground Joint
Click to view available options
Cone Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Standard Taper Joint No.:
NS 19/26
NS 24/29
NS 29/32
NS 45/40
Unit Size:
Each
Description
- Acc. to DIN 12257
- High taper accuracy of the cones
- Minimized need for grease
- Excellent tightness
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Certifications/Compliance | DIN 12257 and in addition to DIN standard |
| Cone Size | NS 29/32 |
| Material | Duran Glass |
| Product Type | Expansion Adapter |
| Quantity | 1 Ea. |
| Standard Taper Joint No. | NS 45/40 |