Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric)Top:
35 mm
45 mm
55 mm
70 mm
80 mm
100 mm
120 mm
150 mm
180 mm
200 mm
300 mm
Unit Size:
Each
Pack of 10
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With short stem
- Due to its shape, it is suitable for liquid-solid filtration using a filter paper
- Fired-on, highly durable white print
Specifications
Specifications
| Diameter (Metric)Top | 150 mm |
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| For Use With (Application) | Decanting and filtration of liquids; The spillage of substances is avoided; Filtering |
| Quantity | 10/Pk. |