Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Expansion from sleeve to core.
Brand:  DWK Life Sciences 286548407
 View more versions of this product
Product Code. 10707101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
  • Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Socket NS 29/32, Cone NS 34/35
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone > Socket NS 29/32, cone NS 34/35