Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
Product Code. 10652351
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
Standard Taper Joint:
NS19/26
NS29/32
Unit Size:
Each
Pack of 10
Product Code. 10652351
Brand: Lenz Laborglasinstrumente™ 03022928
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Spécification
Certifications/Compliance DIN EN ISO 4797
Capacity (Metric) 50 mL
Material Borosilicate Glass, Duran Glass
Includes With scale (approx. volume/mL), with inscription label
Standard Taper Joint NS29/32
Diameter (Metric) Outer 51 mm