Learn More
DWK Life Sciences DURAN™ Erlenmeyer Flask, narrow neck
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
125 mL
150 mL
200 mL
250 mL
300 mL
500 mL
800 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 10
Pack of 2
Description
Thanks to their exceptional properties, such as excellent chemical resistance, nearly inert behaviour and high usage temperature, the original DURAN™ Erlenmeyer flasks are ideal for use in laboratories. Moreover, the uniform wall thickness, minimal thermal expansion and the resultant high thermal shock resistance are outstanding characteristics. Therefore, DURAN™ Erlenmeyer flasks are particularly suited to use with very high temperatures and fast temperature changes.
- Easy mixing due to conical form
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Diameter (Metric) Base | 67 mm |
| Capacity (Metric) | 125 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Media preparation and cultivation; Storing and transport; Mixing several components; Heating; Diluting solutions; Measuring liquid amounts; Size 25 mL without Retrace Code und Sizes 150, 200 and 300 mL non-DIN ISO size |
| No. per Pack | 10/Pk. |
| Autoclavable | Autoclavable |