Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel are manufactured from durable DURAN™ borosilicate glass.
€ 55.00 - € 669.00
Specifications
Material Borosilicate Glass, Duran Glass
Bottom Shape Round
View More Specs
Products 14
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
10720992
View Documents
Lenz Laborglasinstrumente™
03305488
10 L 29/32 NS Side Neck, 45/40 NS Center Neck
€ 669.00
Each
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
10413121
View Documents
Lenz Laborglasinstrumente™
03304470
1000 mL 29/32 NS Side Neck, 29/32 NS Center Neck
€ 138.00
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
10102001
View Documents
Lenz Laborglasinstrumente™
03305470
1000 mL 29/32 NS Side Neck, 45/40 NS Center Neck
€ 101.00
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
10599421
View Documents
Lenz Laborglasinstrumente™
03304137
100 mL 14/23 NS Side Neck, 29/32 NS Center Neck
€ 55.00
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
10662852
View Documents
Lenz Laborglasinstrumente™
03304273
2000 mL 19/26 NS Side Neck, 29/32 NS Center Neck
€ 136.00
Each
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
10070331
View Documents
Lenz Laborglasinstrumente™
03305473
2000 mL 29/32 NS Side Neck, 45/40 NS Center Neck
€ 156.00
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
10612931
View Documents
Lenz Laborglasinstrumente™
03304149
250 mL 14/23 NS Side Neck, 29/32 NS Center Neck
€ 69.00
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
10325831
View Documents
Lenz Laborglasinstrumente™
03304437
100 mL 29/32 NS Side Neck, 29/32 NS Center Neck
€ 108.00
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
10344931
View Documents
Lenz Laborglasinstrumente™
03304249
250 mL 19/26 NS Side Neck, 29/32 NS Center Neck
€ 85.00
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
10261311
View Documents
Lenz Laborglasinstrumente™
03305479
4 L 29/32 NS Side Neck, 45/40 NS Center Neck
€ 298.00
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
10422931
View Documents
Lenz Laborglasinstrumente™
03304158
500 mL 14/23 NS Side Neck, 29/32 NS Center Neck
€ 69.00
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
10519961
View Documents
Lenz Laborglasinstrumente™
03304258
500 mL 19/26 NS Side Neck, 29/32 NS Center Neck
€ 72.00
Each
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
10578081
View Documents
Lenz Laborglasinstrumente™
03305485
6 L 29/32 NS Side Neck, 45/40 NS Center Neck
€ 386.00
Each
Estimated Shipment: 01-04-2025
to see stock.
Add to basket
10273151
View Documents
Lenz Laborglasinstrumente™
03304458
500 mL 29/32 NS Side Neck, 29/32 NS Center Neck
€ 106.00
Each
Estimated Shipment: 17-03-2025
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12392
  • Two parallel side necks at 90° angle
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Borosilicate Glass, Duran Glass
Round