Learn More
Lenz Laborglasinstrumente™ Duran Allihn Condensers with Glass Olives
Product Code. 11896134
Click to view available options
Material:
Duran
Unit Size:
Each
Product Code. 11896134
Brand: Lenz Laborglasinstrumente™ 05223404
Only null left
Add to basket
Add to basket
Specifications
For Use With (Application) For condensing vapors
Material Duran
Type Allihn Condenser
Features With NS socket and NS cone and Glass olives water connection