Learn More
Macherey-Nagel™ Duotest™ pH Paper Refill
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Refills give the user the best possible accuracy in pH measurement.
Brand:  Macherey-Nagel™ 90315
 View more versions of this product
Product Code. 10499860
  • € 34.45 / Pack of 3
Explore more special offers
Description
Description
  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colors for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values
Specifications
Specifications
7 to 10
Refill
3 Pack
pH Paper