Learn More
Euro Protection™ Dunlop™ Hygrade Boots
Product Code. p-8008041
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
Unit Size:
Each
Product Code. 12004475
Brand: Euro Protection™ 9HYGR41
Only null left
Specifications
Certifications/Compliance CE Certified, EN ISO 20347
Color White, Green
Footbed Material PVC
For Use With (Application) For industrial use, food companies and agricultural work
Liner Material Nylon
Material PVC
Size 41
Sole Features Anti-slip
Sole Material PVC
Style Non-safety
Resistant To Water
Waterproof Yes
Product Type Boots