Learn More
Vitlab™ Drying Rack
Product Code. 10274121
Click to view available options
:
Length: 450mm
Product Code. 10274121
Brand: Vitlab™ 76299
Only null left
Add to basket
Add to basket
Vitlab™ Drying Rack with a back plate and trough constructed of PVC and drainage nozzle.
  • Back plate and trough made from PVC with drainage nozzle
  • With 75 metal pegs with PE coating for hanging various sizes of apparatus
  • With two bore holes for simple wall mounting
Specifications
Length (Metric) 450 mm
Height (Metric) 630 mm