Learn More
Witeg™ Dropping Funnel with Pressure Equalization
Use this funnel to slowly add air-sensitive reagents in a sealed environment. Witeg™ Dropping Funnel with Pressure Equalization facilitates the drop-wise addition of reagents.
€ 80.85
Specifications
Material Glass
Description Pressure equalizing
Graduated Yes
Graduations 2mL
View More Specs
Products 1
Product Code Brand Capacity (Metric) Graduations Stopcock Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Stopcock Price Quantity & Availability  
10688115
View Documents
Witeg™
1 866 100
100 mL 2mL ST-PTFE 29/32
€ 80.85
Each
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
Description
Description
  • Pressure equalizing.
  • Graduated.
  • Pack: 2 pieces.
Specifications
Specifications
Glass
Pressure equalizing
Yes
2mL
Documents
Product Certifications
Product Content Correction
Product Title
Witeg™ Dropping Funnel with Pressure Equalization