Learn More
Dräger™ Short-Term Detector Tubes
Product Code. p-307909
Click to view available options
Measurement Range:
0.2 to 2%
40 to 800
Unit Size:
5 tests
Pack of 10
Product Code. 10454292
Brand: Dräger™ 8101511
Specifications
For Use With (Equipment) Accuro
Measurement Range 0.2 to 2%
Pressure Range 1013hpa
Temperature Range (English) 68°F to 104°F
Temperature Range (Metric) 20° to 40°C