Learn More
Dräger™ CMS Chips: Mercaptan
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406360
Product Code. 12314629
  • € 178.00 / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
70 to 480 sec.
0°C to 40°C
Mercaptan CMS Chip
0.25 to 6
32°F to 104°F