Learn More
Dräger™ CMS Chips: Acetone
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406470
Product Code. 12384599
  • € 126.00 / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
0 to 30mg/L
60 to 600 sec.
5° to 40°C
Acetone CMS Chip
40 to 600
41°F to 104°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers