missing translation for 'onlineSavingsMsg'
Learn More

Doxercalciferol-d3, MedChemExpress

Product Code. p-200098969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16451499

Brand: Medchem Express HY15285

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxercalciferol-D3 is the deuterated form of Doxercalciferol, which is a Vitamin D2 analog that acts as a vitamin D receptor activator (VDRA).

Doxercalciferol-D3 is the deuterated form of Doxercalciferol, which is a Vitamin D2 analog that acts as a vitamin D receptor activator (VDRA).

Specifications

Chemical Name or Material Doxercalciferol-d3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C28H41D3O2
Color White
SMILES O[C@@H](C[C@H](O)C1)C(/C1=C([2H])\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)/C=C/[C@H](C)C(C)C)CC3)=C([2H])\[2H]
Molecular Weight (g/mol) 415.67
Quantity 1 mg
Formula Weight 415.67
Percent Purity 96.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.