Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Product Code. p-4246809
Click to view available options
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Heavy Polymer
Metroseal Green Epoxy
Stainless Steel
Unit Size:
Each
Product Code. 15906991
Brand: Metro™ LBTM21S
Only null left
Add to basket
Add to basket
Kit provides components for shelving units that fit on tracks for added mobility
  • For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
  • Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Product Type Mobile Unit Kit
Finish Stainless Steel
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system
Includes Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Width (English) Shelf 21 in.
Width (English) 21 in.
Width (Metric) Shelf 53 cm
Width (Metric) 53 cm
Material Stainless Steel