Learn More
Lenz Laborglasintrumente Dosing Funnel
Product Code. p-7222547
Click to view available options
Capacity (Metric):
1 L
4 L
Unit Size:
Each
Product Code. 11903611
Brand: Lenz Laborglasintrumente 06350079
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • System contents and spare parts for one-way reaction/distillation units 6, 10, 20 l, LF 150
Specifications
Capacity (Metric) 4 L
For Use With (Equipment) 20 L reaction vessel/distillation unit
Joint Number NS 29/32