Learn More
Thermo Scientific™ Domed and Flat PCR Cap Strips
Product Code. p-4525209
Click to view available options
Quantity:
120/Cs.
250/Cs.
Color:
Clear
Unit Size:
Pack of 120
Pack of 250
Product Code. 10690174
Brand: Thermo Scientific™ AB0783
Only null left
Add to basket
Add to basket
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polypropylene
No. of Wells 8
Quantity 120/Cs.
Format Strips of 8 Flat Caps
Product Type PCR Plate Cap Strip
Certified RNase/DNase-free Yes
Plate Compatibility 96-well Plate