missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4E6H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66758-1-IG-150UL
This item is not returnable.
View return policy
Description
DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.Specifications
| DKK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QUN9, Q9UBP4 | |
| DKK3 | |
| DKK3 Fusion Protein Ag23615 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 4E6H6 | |
| Unconjugated | |
| DKK3 | |
| Dickkopf 3, Dickkopf related protein 3, Dkk 3, DKK3, hDkk 3, REIC, RIG | |
| Mouse | |
| Protein A | |
| RUO | |
| 171548, 27122, 50781, 664653 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction