missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Distillation Standard, Paragon Scientific™
Click to view available options
Quantity:
250 mL
Description
- Manufactured in accordance with ASTM / EN or IP test method protocol
- ISO 17034 fully certified products available
- Tested exclusively by ISO 17025 accredited laboratories for ISO 17034 CRM's
- Ensures good interlaboratory correlation
- Supplied in tamper evident UN compliant security pack
- Pack size 250mL
- 12 months shelf life
Specifications
Specifications
Quantity | 250 mL |
Safety and Handling
Product Identifier
- CRMU-DIGO
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Aspiration toxicity Category 1
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
Nominal values quoted
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'