Learn More
Contec™ Disposal Bag for Mop Removal Tool
Disposal bag for mop head removal tool.
Brand: Contec™ BNBG0001
224.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Broad spectrum efficacy in 3 mins saves time spent on biodecontamination.
- Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
- Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
- No hazard classification so only basic PPE required and no special disposal is necessary
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom — this needs a bit of random text deleting from the end
Spécification
Mop Removal Tool | |
Not Autoclavable | |
Single-use | |
Non-sterile |
LDPE | |
Disposal bag for mop head removal tool | |
80 Bags/Cs. |
Suggestions de produits
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
013890 | Certificate of Compliance | 03/05/2021 | BNBG0001 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.