missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Source™ Disposable Chemical Resistant SMS Coveralls Type 5/6
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
X-Large
Description
- Conforms to CE Category III Type 5/6
- Non-woven SMS disposable fabric
- Protects against chemical and biological hazards
Specifications
Specifications
Size | Large |
Gender | Unisex |
Material | Non-woven SMS |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Flap |
Seam Type | Stitched |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'