Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
Brand: 3M™ 7100003010
This item is not returnable.
View return policy
Description
3M Particulate Respirator P1 P1 for mechanically generated particles grinding, sanding, crushing and sawing Not TGA Approved Not allowed for Clinical Use
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.Specifications
European Community Directive 89/686/EEC, EN 149:2001 A1:2009, CE | |
Single-use | |
Particulate Respirator | |
Protection against dust and mist | |
8810 | |
Dusts, mists and Metal fumes |
Twin strap | |
FFP2 NR D | |
No | |
Yes | |
1 Ea. | |
Convex |