Learn More
Disposable Luer-Slip Two-Part Tuberculin Syringe
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 100
Description
Includes
Tuberculin Syringes only. (Needles not included; sold separately.)
- Fully visible glass barrel for visual verification
- Ergonomic light weight metal, plastic design
- 2mlL barrel with 0.1mL increment trigger
- Safe and easy needle change out via luer lock connection
- Limited spare parts for easy service
- Boilable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | DIN, EN, ISO and FDA standards, Quality management system certified according to ISO 13485:2003 / AC 2007, CE-marked |
| Material | Polypropylene |
| Color | Transparent Barrel |
| Description | Smudge-proof scale |
| Includes | Syringes only. (Needles are sold separately.) |
| Latex Free | Yes |
| Product Type | Luer-slip 2-part Tuberculin Syringe |
| Disposable | Single-use |
| Graduations | 0.01 mL |
| Show More |