missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 40 g/m2
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Light and breathable polypropylene material
- Loose shape allows user to maintain good movement
- Distribution of fibers guarantees opacity
- Collarless shirt
- Pants with elasticated waist
- Individual packaging for hygiene and controlled distribution
- 40 g/m2 fabric weight
Specifications
Specifications
Size | X-Large |
Material | SMS |
Color | Blue |
Sleeve Style | Short Sleeve |
Disposable | Single-use |
For Use With (Application) | Dental, Medical |
No. per Pack | 50 Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'