Learn More
Honeywell™ Dispensing System and Dip Tube
Product Code. 16891210 Shop All Honeywell Fluka Products
Click to view available options
For Use With (Equipment):
18 L Drum without Dip Tube
200 L Drum without Dip Tube
45 L Drum without Dip Tube
Capacity (Metric):
18 L
45 L
200 L
Unit Size:
1 pack
Product Code. 16891210
Brand: Honeywell™ 89889H1EA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dispensing system and dip tube
TRUSTED_SUSTAINABILITY
Specifications
Product Type Returnable Container Accessory
Capacity (Metric) 200 L
For Use With (Equipment) 200 L Drum without Dip Tube
Quantity 1
Weight (Metric) 1.22 Kg